Learn More
Alfa Aesar™ 2-Chloro-N-(3-chloro-4-methylphenyl)benzamide, 97%
Brand:  Alfa Aesar™ H59829.03
Additional Details : CAS Number : 7017-26-7 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15486248
  • 6228.40 SEK / 1g
Estimated Shipment date
from Supplier 05-07-2018
Add to basket
Specifications
Chemical Identifiers
2-Chloro-N-(3-chloro-4-methylphenyl)benzamide
7017-26-7
280.15
MFCD00090964
Specifications
2-Chloro-N-(3-chloro-4-methylphenyl)benzamide
1g
Solid
280.15
7017-26-7
Yellow-brown
97%
MFCD00090964