Läs mer
2-Chloro-N-phenylacetamide, TRC
Produktkod. 30361668
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30361668 25 g
1 options
Produktkod. 30361668 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37537025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Chloro-N-phenylacetamide
CAS 587-65-5
Molekylformel C8H8ClNO
Kvantitet 25 g
Synonym 2-Chloroacetanilide, Chloracetanilide, Chloroacetanilide, N-(2-Chloroacetyl)aniline, N-(Chloroacetyl)aniline, N-Phenyl-2-chloroacetamide, N-Phenyl-2-chloroethanamide, N-Phenylchloroacetamide, NSC 311, Phenylcarbamoylmethyl Chloride, alpha-Chloro-N-phenylacetamide, alpha-Chloroacetanilide, ω-Chloroacetanilide
InChI formel InChI=1S/C8H8ClNO/c9-6-8(11)10-7-4-2-1-3-5-7/h1-5H,6H2,(H,10,11)
LEDER ClCC(=O)Nc1ccccc1
IUPAC-namn 2-chloro-N-phenylacetamide
Molekylvikt (g/mol) 169.03
Formel vikt 169.0294