Läs mer
2-Chloroadenosine 5’-Monophosphate Ditriethylamine Salt, TRC
Produktkod. 30364787
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364787 10 mg
30353659 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364787 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36410010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52621-99-5
Molekylformel C10 H13 Cl N5 O7 P . 2 C6 H15 N
Molekylvikt (g/mol) 584.046
Synonym 2-Chloro-5'-adenylic Acid, 2-Chloro-adenosine 5'-(Dihydrogen Phosphate) Ditriethylamine, 2-Chloro-AMP Ditriethylamine
IUPAC-namn [(2R,3S,4R,5R)-5-(6-amino-2-chloropurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;N,N-diethylethanamine
LEDER CCN(CC)CC.CCN(CC)CC.Nc1nc(Cl)nc2c1ncn2[C@@H]3O[C@H](COP(=O)(O)O)[C@@H](O)[C@H]3O
Specifikationer
Kemiskt namn eller material 2-Chloroadenosine 5’-Monophosphate Ditriethylamine Salt
Kvantitet 10 mg
InChI formel InChI=1S/C10H13ClN5O7P.2C6H15N/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(18)5(17)3(23-9)1-22-24(19,20)21;2*1-4-7(5-2)6-3/h2-3,5-6,9,17-18H,1H2,(H2,12,14,15)(H2,19,20,21);2*4-6H2,1-3H3/t3-,5-,6-,9-;;/m1../s1
Formel vikt 583.265