missing translation for 'onlineSavingsMsg'
Läs mer

2-Chlorobenzyl-3,4,5,6-d4 Chloride, TRC

Produktkod. 30359891
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359891 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30359891 Supplier Toronto Research Chemicals Supplier No. TRCC364227250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-Chlorobenzyl-3,4,5,6-d4 Chloride
CAS 2732862-20-1
Molekylformel C7 D4 H2 Cl2
Kvantitet 250 mg
Synonym 2-Chlorobenzyl-3,4,5,6-d4 Chloride, 2-Chlorobenzyl Chloride-3,4,5,6-D4, 1-Chloro-2-(chloromethyl)benzene-3,4,5,6-d4, 5-Chloro-6-(chloromethyl)benzene-1,2,3,4-d4
InChI formel InChI=1S/C7H6Cl2/c8-5-6-3-1-2-4-7(6)9/h1-4H,5H2/i1D,2D,3D,4D
LEDER [2H]c1c([2H])c([2H])c(CCl)c(Cl)c1[2H]
IUPAC-namn 1-chloro-2-(chloromethyl)-3,4,5,6-tetradeuteriobenzene
Molekylvikt (g/mol) 165.0532
Formel vikt 164.0098
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.