missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-86646
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1622-32-8
C2H4Cl2O2S
163.02
MFCD00007461
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
15385
2-chloroethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl
This item is not returnable. View return policy
1622-32-8
C2H4Cl2O2S
163.02
MFCD00007461
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
15385
2-chloroethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.5500g/mL
Boiling Point 78°C (9.0 mmHg)
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula ClCH2CH2SO2Cl
Refractive Index 1.4910 to 1.494
Quantity 5 g
Specific Gravity 1.55
Formula Weight 163.02
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanesulfonyl chloride, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'