missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7022547
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
This item is not returnable. View return policy
1622-32-8
C2H4Cl2O2S
163.012
MFCD00007461
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
15385
2-chloroethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl
This item is not returnable. View return policy
1622-32-8
C2H4Cl2O2S
163.012
MFCD00007461
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
15385
2-chloroethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl

2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
TRUSTED_SUSTAINABILITY
Density 1.56
Boiling Point 203°C
Flash Point >110°C (230°F)
Linear Formula ClCH2CH2SO2Cl
Refractive Index 1.4915
Quantity 10 g
UN Number UN3390
Beilstein 1751202
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 163.02
Percent Purity 98%
Chemical Name or Material 2-Chloroethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'