Läs mer
2-Chloroethyl 4-methylbenzenesulfonate, TRC
Produktkod. 30355450
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355450 1 g
30350936 5 g
30357789 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355450 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3632151G
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80-41-1
Molekylformel C9H11ClO3S
Molekylvikt (g/mol) 234.7
Synonym 2-(p-Toluenesulfonyloxy)ethyl chloride, 2-Chloro-1-(p-toluenesulfonyl)ethane, 2-Chloroethanol p-toluenesulfonate, 2-Chloroethanol tosylate, 2-Chloroethyl p-toluenesulfonate, 2-Chloroethyl p-tosylate, 2-Chloroethyl tosylate, 2-Tosyloxyethyl chloride, p-Toluenesulfonic acid 2-chloroethyl ester, beta-Chloroethyl p-toluenesulfonate, beta-Chloroethyl tosylate
IUPAC-namn 2-chloroethyl 4-methylbenzenesulfonate
LEDER Cc1ccc(cc1)S(=O)(=O)OCCCl
Specifikationer
Kemiskt namn eller material 2-Chloroethyl 4-methylbenzenesulfonate
Kvantitet 1 g
InChI formel InChI=1S/C9H11ClO3S/c1-8-2-4-9(5-3-8)14(11,12)13-7-6-10/h2-5H,6-7H2,1H3
Formel vikt 234.0117