Läs mer
2-Chloroethylphosphoryl Dichloride, TRC
Produktkod. 30368721
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368721 250 mg
30367619 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368721 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC984690250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Chloroethylphosphoryl Dichloride
CAS 1455-05-6
Molekylformel C2H4O2PCl3
Kvantitet 250 mg
InChI formel InChI=1S/C2H4Cl3O2P/c3-1-2-7-8(4,5)6/h1-2H2
LEDER ClCCOP(=O)(Cl)Cl
IUPAC-namn 1-chloro-2-dichlorophosphoryloxyethane
Molekylvikt (g/mol) 197.38
Formel vikt 195.9014