Läs mer
2-Chloroimidazo[1,2-a]pyridine-6-carbonitrile, TRC
Produktkod. 30349122
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30349122 5 mg -
30360793 25 mg -
30353666 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349122 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3829305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1019020-03-1
Molekylformel C8H4N3Cl
Molekylvikt (g/mol) 177.59
IUPAC-namn 2-chloroimidazo[1,2-a]pyridine-6-carbonitrile
LEDER Clc1cn2cc(ccc2n1)C#N
Specifikationer
Kemiskt namn eller material 2-Chloroimidazo[1,2-a]pyridine-6-carbonitrile
Kvantitet 5 mg
InChI formel InChI=1S/C8H4ClN3/c9-7-5-12-4-6(3-10)1-2-8(12)11-7/h1-2,4-5H
Formel vikt 177.0094