missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-1,3-benzoxazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7077419
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
41014-43-1
C8H6ClNO
167.59
MFCD05664964
ANRDUCQCZKLSGF-UHFFFAOYSA-N
2-chloromethyl-1,3-benzoxazole, 2-chloromethyl benzo d oxazole, 2-chloromethyl benzoxazole, benzoxazole, 2-chloromethyl, chloromethylbenzoxazole, 2-chloromethylbenzoxazole, 2-chloromethyl-benzooxazole, chloromethyl-benzoxazole, acmc-1aqt5
2061989
2-(chloromethyl)-1,3-benzoxazole
ClCC1=NC2=CC=CC=C2O1
This item is not returnable. View return policy
41014-43-1
C8H6ClNO
167.59
MFCD05664964
ANRDUCQCZKLSGF-UHFFFAOYSA-N
2-chloromethyl-1,3-benzoxazole, 2-chloromethyl benzo d oxazole, 2-chloromethyl benzoxazole, benzoxazole, 2-chloromethyl, chloromethylbenzoxazole, 2-chloromethylbenzoxazole, 2-chloromethyl-benzooxazole, chloromethyl-benzoxazole, acmc-1aqt5
2061989
2-(chloromethyl)-1,3-benzoxazole
ClCC1=NC2=CC=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 80.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)-1,3-benzoxazole
compliance-icons
Product Identifier
  • 2-(Chloromethyl)-1,3-benzoxazole, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'