Läs mer
2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride, Mikromol™
Produktkod. 30050162
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050162 25 mg
18042070 100 mg
30463322 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30050162 Leverantör Mikromol Leverantörsnummer MM0618.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
CAS 153259-31-5
Analyt- eller komponentnamn 2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
Frakt skick Room Temperature
Molekylformel C11 H16 Cl N O2 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym 2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
InChI formel InChI=1S/C11H16ClNO2.ClH/c1-9-10(8-12)13-5-4-11(9)15-7-3-6-14-2;/h4-5H,3,6-8H2,1-2H3;1H
LEDER Cl.COCCCOc1ccnc(CCl)c1C
IUPAC-namn 2-(chloromethyl)-4-(3-methoxypropoxy)-3-methylpyridine;hydrochloride
Molekylvikt (g/mol) 266.16
Kvantitet 25 mg
Formel vikt 265.0636 g/mol