Läs mer
2-Chloromethyl-3H-benzoimidazole-5-carbonitrile, TRC
Produktkod. 30366041
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366041 10 mg
30357018 100 mg
30356788 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366041 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC59810310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 150613-50-6
Molekylformel C9H6N3Cl
Molekylvikt (g/mol) 191.61
IUPAC-namn 2-(chloromethyl)-3H-benzimidazole-5-carbonitrile
LEDER ClCc1nc2ccc(cc2[nH]1)C#N
Specifikationer
Kemiskt namn eller material 2-Chloromethyl-3H-benzoimidazole-5-carbonitrile
Kvantitet 10 mg
InChI formel InChI=1S/C9H6ClN3/c10-4-9-12-7-2-1-6(5-11)3-8(7)13-9/h1-3H,4H2,(H,12,13)
Formel vikt 191.025