missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole, 97%, Thermo Scientific™

Catalog Number p-4228519
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33575-83-6
C9H7ClN2O
194.62
MFCD00466332
AGLNTFQAHIRTFA-UHFFFAOYSA-N
2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene
314941
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
ClCC1=NN=C(O1)C1=CC=CC=C1
This item is not returnable. View return policy
33575-83-6
C9H7ClN2O
194.62
MFCD00466332
AGLNTFQAHIRTFA-UHFFFAOYSA-N
2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene
314941
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
ClCC1=NN=C(O1)C1=CC=CC=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 119.3°C to 119.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 194.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(chloromethyl)-5-phenyl-1, 3, 4-oxadiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'