Läs mer
2-(Chloromethyl)benzaldehyde, TRC
Produktkod. 30362223
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362223 50 mg
30356248 25 mg
30352277 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362223 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36575050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 108683-62-1
Molekylformel C8H7ClO
Molekylvikt (g/mol) 154.59
IUPAC-namn 2-(chloromethyl)benzaldehyde
LEDER ClCc1ccccc1C=O
Specifikationer
Kemiskt namn eller material 2-(Chloromethyl)benzaldehyde
Kvantitet 50 mg
InChI formel InChI=1S/C8H7ClO/c9-5-7-3-1-2-4-8(7)6-10/h1-4,6H,5H2
Formel vikt 154.0185