missing translation for 'onlineSavingsMsg'
Läs mer

(2-Chlorophenyl)diphenylmethyl Ethyl Ether, Mikromol™

Product Code. 30051605
Change view
Click to view available options
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30051605 25 mg
30050439 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051605 Supplier Mikromol Supplier No. MM0015.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2-Chlorophenyl)diphenylmethyl Ethyl Ether
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (2-Chlorophenyl)diphenylmethyl EthylEther
CAS 717109-43-8
Analyt- eller komponentnamn (2-Chlorophenyl)diphenylmethyl EthylEther
Frakt skick Room Temperature
Molekylformel C21 H19 Cl O
Certifikat och efterlevnad ISO 9001
Synonym (2-Chlorophenyl)diphenylmethyl EthylEther
InChI formel InChI=1S/C21H19ClO/c1-2-23-21(17-11-5-3-6-12-17,18-13-7-4-8-14-18)19-15-9-10-16-20(19)22/h3-16H,2H2,1H3
LEDER CCOC(c1ccccc1)(c2ccccc2)c3ccccc3Cl
IUPAC-namn 1-chloro-2-[ethoxy(diphenyl)methyl]benzene
Molekylvikt (g/mol) 322.83
Kvantitet 25 mg
Formel vikt 322.1124 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.