Läs mer
2-Chloroquinolin-6-ol, TRC
Produktkod. 30351345
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351345 500 mg
30368536 100 mg
30357367 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30351345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC429278500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 577967-89-6
Molekylformel C9 H6 Cl N O
Molekylvikt (g/mol) 179.6
Synonym 2-Chloro-6-quinolinol, 2-Chloro-6-hydroxyquinoline
IUPAC-namn 2-chloroquinolin-6-ol
LEDER Oc1ccc2nc(Cl)ccc2c1
Specifikationer
Kemiskt namn eller material 2-Chloroquinolin-6-ol
Kvantitet 500 mg
InChI formel InChI=1S/C9H6ClNO/c10-9-4-1-6-5-7(12)2-3-8(6)11-9/h1-5,12H
Formel vikt 179.0138