Läs mer
2-Chloroquinoline-6-sulfonyl Chloride, TRC
Produktkod. 30361345
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361345 250 mg
30358299 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC381240250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 205055-71-6
Molekylformel C9 H5 Cl2 N O2 S
Molekylvikt (g/mol) 262.11
IUPAC-namn 2-chloroquinoline-6-sulfonyl chloride
LEDER Clc1ccc2cc(ccc2n1)S(=O)(=O)Cl
Specifikationer
Kemiskt namn eller material 2-Chloroquinoline-6-sulfonyl Chloride
Kvantitet 250 mg
InChI formel InChI=1S/C9H5Cl2NO2S/c10-9-4-1-6-5-7(15(11,13)14)2-3-8(6)12-9/h1-5H
Formel vikt 260.9418