missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroquinoline, 99%, Thermo Scientific™

Catalog Number p-7027010
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
612-62-4
C9H6ClN
163.604
MFCD00006741
OFUFXTHGZWIDDB-UHFFFAOYSA-N
2-chloro-quinoline, quinoline, 2-chloro, chloroquinoline, unii-ux7rin8gew, quinoline, chloro, ccris 3977, ux7rin8gew, chioroquinoline, 2-chloro quinoline, pubchem7580
11928
2-chloroquinoline
C1=CC=C2C(=C1)C=CC(=N2)Cl
This item is not returnable. View return policy
612-62-4
C9H6ClN
163.604
MFCD00006741
OFUFXTHGZWIDDB-UHFFFAOYSA-N
2-chloro-quinoline, quinoline, 2-chloro, chloroquinoline, unii-ux7rin8gew, quinoline, chloro, ccris 3977, ux7rin8gew, chioroquinoline, 2-chloro quinoline, pubchem7580
11928
2-chloroquinoline
C1=CC=C2C(=C1)C=CC(=N2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloroquinoline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It possess antimicrobial activity against a panel of bacterial and fungal strains. It reacts with benzotriazole, followed by cyclization with loss of nitrogen, gives the indolo[2,3-b]quinoline ring system for use in novel cytotoxic DNA topoisomerase II inhibitors.

Solubility
Soluble in ethanol, ether, methanol, and chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Melting Point 34°C to 37°C
Density 1.23
Boiling Point 265°C to 267°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 5 g
Beilstein 112561
Solubility Information Soluble in ethanol,ether,methanol,and chloroform. Insoluble in water.
Formula Weight 163.61
Percent Purity 99%
Chemical Name or Material 2-Chloroquinoline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'