Läs mer
2-Cl-IB-MECA, TRC
Produktkod. 30362873
Change view
Kvantitet:
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30362873 2.5 mg 2.5mg
30356699 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30362873 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5762302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163042-96-4
Molekylformel C18 H18 Cl I N6 O4
Molekylvikt (g/mol) 544.73
IUPAC-namn (2S,3S,4R,5R)-5-[2-chloro-6-[(3-iodophenyl)methylamino]purin-9-yl]-3,4-dihydroxy-N-methyloxolane-2-carboxamide
LEDER CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NCc4cccc(I)c4)nc(Cl)nc23
Specifikationer
Kemiskt namn eller material 2-Cl-IB-MECA
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C18H18ClIN6O4/c1-21-16(29)13-11(27)12(28)17(30-13)26-7-23-10-14(24-18(19)25-15(10)26)22-6-8-3-2-4-9(20)5-8/h2-5,7,11-13,17,27-28H,6H2,1H3,(H,21,29)(H,22,24,25)/t11-,12+,13-,17+/m0/s1
Formel vikt 544.0123
Procent renhet >95%