Läs mer
2-Cyano-2-cyclohexylideneacetic Acid Ethyl Ester, TRC
Produktkod. 30365780
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30365780 500 mg
1 options
Produktkod. 30365780 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC962600500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Cyano-2-cyclohexylideneacetic Acid Ethyl Ester
CAS 6802-76-2
Molekylformel C11H15NO2
Kvantitet 500 mg
Synonym Ethyl Cyanocyclohexylideneacetate, NSC 10726
InChI formel InChI=1S/C11H15NO2/c1-2-14-11(13)10(8-12)9-6-4-3-5-7-9/h2-7H2,1H3
LEDER CCOC(=O)C(=C1CCCCC1)C#N
IUPAC-namn ethyl 2-cyano-2-cyclohexylideneacetate
Molekylvikt (g/mol) 193.24
Formel vikt 193.1103