Läs mer
2-Cyano-3-hydroxypyridine, TRC
Produktkod. 30357383
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357383 500 mg
30356784 250 mg
30354615 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30357383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC987963500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 932-35-4
Molekylformel C6H4N2O
Molekylvikt (g/mol) 120.11
Synonym 2-Cyano-3-pyridinol, 3-Hydroxy-2-cyanopyridine, 3-Hydroxypicolinonitrile, 3-Hydroxy-2-pyridinecarbonitrile, 3-Hydroxy-(7CI,8CI)picolinonitrile
IUPAC-namn 3-hydroxypyridine-2-carbonitrile
LEDER Oc1cccnc1C#N
Specifikationer
Kemiskt namn eller material 2-Cyano-3-hydroxypyridine
Kvantitet 500 mg
InChI formel InChI=1S/C6H4N2O/c7-4-5-6(9)2-1-3-8-5/h1-3,9H
Formel vikt 120.0324