missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals

Catalog Number p-7025658
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
114772-53-1
C14H11N
193.25
MFCD00151805
ZGQVZLSNEBEHFN-UHFFFAOYSA-N
2-cyano-4'-methylbiphenyl, 4'-methyl-2-cyanobiphenyl, 4'-methylbiphenyl-2-carbonitrile, 2'-cyano-4-methylbiphenyl, 2-p-tolyl benzonitrile, 2-4-methylphenyl benzonitrile, 2-cyano-4'-methyl biphenyl, 4'-methyl-1,1'-biphenyl-2-carbonitrile, unii-o417ti15ta, 4'-methyl-2-cyano diphenyl
145512
2-(4-methylphenyl)benzonitrile
CC1=CC=C(C=C1)C1=CC=CC=C1C#N
This item is not returnable. View return policy
114772-53-1
C14H11N
193.25
MFCD00151805
ZGQVZLSNEBEHFN-UHFFFAOYSA-N
2-cyano-4'-methylbiphenyl, 4'-methyl-2-cyanobiphenyl, 4'-methylbiphenyl-2-carbonitrile, 2'-cyano-4-methylbiphenyl, 2-p-tolyl benzonitrile, 2-4-methylphenyl benzonitrile, 2-cyano-4'-methyl biphenyl, 4'-methyl-1,1'-biphenyl-2-carbonitrile, unii-o417ti15ta, 4'-methyl-2-cyano diphenyl
145512
2-(4-methylphenyl)benzonitrile
CC1=CC=C(C=C1)C1=CC=CC=C1C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 49°C to 53°C
Density 1.17
Boiling Point >320°C
Quantity 5 g
UN Number UN3077
Beilstein 3605954
Formula Weight 193.26
Percent Purity ≥98%
Chemical Name or Material 2-Cyano-4'-methylbiphenyl
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'