missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-fluoropyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-7030066
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
3939-15-9
C6H3FN2
122.10
MFCD09263323
NVOLTPVZQXTZCW-UHFFFAOYSA-N
2-cyano-6-fluoropyridine, 6-fluoropicolinonitrile, 2-pyridinecarbonitrile, 6-fluoro, 6-fluoro-2-cyanopyridine;2-fluoro-6-cyanopyridine;6-fluoropicolinonitrile, pubchem4017, acmc-1cj0q, 2-pyridinecarbonitrile,6-fluoro, 6-fluoro-pyridine-2-carbonitrile, 2-fluoropyridine-6-carbonitrile, 6-fluoro-2-pyridinecarbonitrile
14296369
6-fluoropyridine-2-carbonitrile
FC1=CC=CC(=N1)C#N
This item is not returnable. View return policy
3939-15-9
C6H3FN2
122.10
MFCD09263323
NVOLTPVZQXTZCW-UHFFFAOYSA-N
2-cyano-6-fluoropyridine, 6-fluoropicolinonitrile, 2-pyridinecarbonitrile, 6-fluoro, 6-fluoro-2-cyanopyridine;2-fluoro-6-cyanopyridine;6-fluoropicolinonitrile, pubchem4017, acmc-1cj0q, 2-pyridinecarbonitrile,6-fluoro, 6-fluoro-pyridine-2-carbonitrile, 2-fluoropyridine-6-carbonitrile, 6-fluoro-2-pyridinecarbonitrile
14296369
6-fluoropyridine-2-carbonitrile
FC1=CC=CC(=N1)C#N

It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 32°C to 35°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 122.1
Percent Purity 95%
Chemical Name or Material 2-Cyano-6-fluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'