Läs mer
2-Cyano-N-methylacetamide, TRC
Produktkod. 30361413
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30361413 5 g
1 options
Produktkod. 30361413 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9818605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Cyano-N-methylacetamide
CAS 6330-25-2
Molekylformel C4H6N2O
Kvantitet 5 g
Synonym 2-Cyano-N-methylacetamide, Cyanoacetic Acid Methylamide, Cyanoacetic Acid Monomethylamide, N-Methyl-2-cyanoacetamide, N-Methylcyanoacetamide, NSC 47005
InChI formel InChI=1S/C4H6N2O/c1-6-4(7)2-3-5/h2H2,1H3,(H,6,7)
LEDER CNC(=O)CC#N
IUPAC-namn 2-cyano-N-methylacetamide
Molekylvikt (g/mol) 98.1
Formel vikt 98.048