Läs mer
2-Cyanoacetohydrazide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Cyanoacetohydrazide |
| CAS | 140-87-4 |
| Molekylformel | C3H5N3O |
| Kvantitet | 1 g |
| Synonym | Cyano-acetic Acid Hydrazide, (Cyanoacetyl)hydrazine, (alpha-Cyanoacetyl)hydrazine, 2-Cyanoacethydrazide, 2-Cyanoacetohydrazide, 2-Cyanoacetylhydrazide, AB-42, Armazal, Cianazil, Cyacetacid, Cyacetacide, Cyacetazid, Cyacetazide, Cyanacethydrazide, Cyanacetic Acid Hydrazide, Cyanacetohydrazide, Cyanacetylhydrazide, Cyanazide, Cyanizide, Cyanoacethydrazide, Cyanoacetic Acid Hydrazide, Cyanoacetic Hydrazide, Cyanoacetohydrazide, Cyanoacetyl Hydrazide, Cyanoethanoic Hydrazide, Cyazid, Cyazide, Dictycide, Dictyzide, Helmox, Hidacian, Hidaciann, Leandin, Mackreazid, Malonitrile Hydrazide, Malononitrile Hydrazide, NSC 24261, Neohydrazid, Reacid, Reazid, Reazide, Tsiazid, alpha-Cyanoacetohydrazide |
| InChI formel | InChI=1S/C3H5N3O/c4-2-1-3(7)6-5/h1,5H2,(H,6,7) |
| LEDER | NNC(=O)CC#N |
| IUPAC-namn | 2-cyanoacetohydrazide |
| Molekylvikt (g/mol) | 99.09 |
| Formel vikt | 99.0433 |