Läs mer
2-Cyanoethyl Phosphate, Barium, TRC
Produktkod. 30360449
Change view
Kvantitet:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360449 5 g
30354540 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360449 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9809505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Cyanoethyl Phosphate, Barium Salt Dihydrate
CAS 5015-38-3
Molekylformel C3 H4 N O4 P . Ba
Kvantitet 5 g
InChI formel InChI=1S/C3H6NO4P.Ba/c4-2-1-3-8-9(5,6)7;/h1,3H2,(H2,5,6,7);/q;+2/p-2
LEDER [Ba+2].[O-]P(=O)([O-])OCCC#N
IUPAC-namn barium(2+);2-cyanoethyl phosphate
Molekylvikt (g/mol) 286.37
Formel vikt 286.893