Läs mer
2-(Cyanomethyl)indole, TRC
Produktkod. 30367258
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367258 25 mg
30363514 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367258 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98192025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7210-27-7
Molekylformel C10 H8 N2
Molekylvikt (g/mol) 156.18
Synonym 1H-Indole-2-acetonitrile
IUPAC-namn 2-(1H-indol-2-yl)acetonitrile
LEDER N#CCc1cc2ccccc2[nH]1
Specifikationer
Kemiskt namn eller material 2-(Cyanomethyl)indole
Kvantitet 25 mg
InChI formel InChI=1S/C10H8N2/c11-6-5-9-7-8-3-1-2-4-10(8)12-9/h1-4,7,12H,5H2
Formel vikt 156.0687