Läs mer
2-Cyclohexylethanol, TRC
Produktkod. 30352041
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30352041 50 g
1 options
Produktkod. 30352041 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99260050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Cyclohexylethanol
CAS 4442-79-9
Molekylformel C8H16O
Kvantitet 50 g
Synonym Cyclohexaneethanol, 2-Cyclohexaneethanol, 2-Cyclohexyl-1-ethanol, Cyclohexylethanol, Cyclohexylethyl alcohol, NSC 30157, beta-Cyclohexylethyl alcohol
InChI formel InChI=1S/C8H16O/c9-7-6-8-4-2-1-3-5-8/h8-9H,1-7H2
LEDER OCCC1CCCCC1
IUPAC-namn 2-cyclohexylethanol
Molekylvikt (g/mol) 128.21
Formel vikt 128.1201