Läs mer
2-Cyclohexylideneacetic Acid, TRC
Produktkod. 30363627
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363627 50 mg
30359158 100 mg
30363525 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363627 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99214850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1552-91-6
Molekylformel C8H12O2
Molekylvikt (g/mol) 140.18
Synonym Cyclohexylideneacetic acid, NSC 59877
IUPAC-namn 2-cyclohexylideneacetic acid
LEDER OC(=O)C=C1CCCCC1
Specifikationer
Kemiskt namn eller material 2-Cyclohexylideneacetic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C8H12O2/c9-8(10)6-7-4-2-1-3-5-7/h6H,1-5H2,(H,9,10)
Formel vikt 140.0837