Läs mer
2-Cyclopropylcarbonyl-3-dimethylaminoacrylic Acid Methyl Ester, TRC
Produktkod. 30363050
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30363050 500 mg
1 options
Produktkod. 30363050 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC988455500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Cyclopropylcarbonyl-3-dimethylaminoacrylic Acid Methyl Ester
CAS 340774-26-7
Molekylformel C10H15NO3
Kvantitet 500 mg
Synonym alpha-[(Dimethylamino)methylene]-beta-oxo-cyclopropanepropanoic Acid Methyl Ester, Methyl 2-(Cyclopropylcarbonyl)-3-(dimethylamino)acrylate, Methyl 2-(Cyclopropylcarbonyl)-3-(dimethylamino)prop-2-enoate
InChI formel InChI=1S/C10H15NO3/c1-11(2)6-8(10(13)14-3)9(12)7-4-5-7/h6-7H,4-5H2,1-3H3/b8-6-
LEDER COC(=O)\C(=C/N(C)C)\C(=O)C1CC1
IUPAC-namn methyl (Z)-2-(cyclopropanecarbonyl)-3-(dimethylamino)prop-2-enoate
Molekylvikt (g/mol) 197.23
Formel vikt 197.1052