Läs mer
2-Demethoxy-4-methoxy Urapidil, TRC
Produktkod. 30133258
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133258 50 mg
30133504 10 mg
30009249 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133258 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23113050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34661-79-5
Molekylformel C20H29N5O3
Molekylvikt (g/mol) 387.48
Synonym 6-[[3-[4-(4-Methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione;
LEDER COc1ccc(cc1)N2CCN(CCCNC3=CC(=O)N(C)C(=O)N3C)CC2
Specifikationer
Kemiskt namn eller material 2-Demethoxy-4-methoxy Urapidil
Kvantitet 50 mg
InChI formel InChI=1S/C20H29N5O3/c1-22-18(15-19(26)23(2)20(22)27)21-9-4-10-24-11-13-25(14-12-24)16-5-7-17(28-3)8-6-16/h5-8,15,21H,4,9-14H2,1-3H3
Formel vikt 387.227