Läs mer
2-Deoxy-2,2-difluoro-4,5-O-isopropylidene-D-threo-pentonic Acid Ethyl Ester Benzoate, TRC
Produktkod. 30356012
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30356012 10 mg
1 options
Produktkod. 30356012 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23282010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Deoxy-2,2-difluoro-4,5-O-isopropylidene-D-threo-pentonic Acid Ethyl Ester Benzoate
CAS 143234-92-8
Molekylformel C17H20F2O6
Kvantitet 10 mg
Synonym 2-Deoxy-2,2-difluoro-4,5-O-(1-methylethylidene)-D-threo-pentonic Acid Ethyl Ester Benzoate
InChI formel InChI=1S/C17H20F2O6/c1-4-22-15(21)17(18,19)13(12-10-23-16(2,3)25-12)24-14(20)11-8-6-5-7-9-11/h5-9,12-13H,4,10H2,1-3H3/t12-,13+/m1/s1
LEDER CCOC(=O)C(F)(F)[C@@H](OC(=O)c1ccccc1)[C@H]2COC(C)(C)O2
IUPAC-namn [(1S)-1-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-3-ethoxy-2,2-difluoro-3-oxopropyl] benzoate
Molekylvikt (g/mol) 358.33
Formel vikt 358.1228