Läs mer
2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibenzoate, TRC
Produktkod. 30367464
Change view
Kvantitet:
1 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367464 1 g
30363141 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367464 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2327751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 122111-01-7
Molekylformel C19 H14 F2 O6
Molekylvikt (g/mol) 376.31
Synonym D-erythro-Pentonic acid, 2-deoxy-2,2-difluoro-, γ-lactone, 3,5-dibenzoate
IUPAC-namn [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
LEDER FC1(F)[C@H](OC(=O)c2ccccc2)[C@@H](COC(=O)c3ccccc3)OC1=O
Specifikationer
Kemiskt namn eller material 2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibe
Kvantitet 1 g
InChI formel InChI=1S/C19H14F2O6/c20-19(21)15(27-17(23)13-9-5-2-6-10-13)14(26-18(19)24)11-25-16(22)12-7-3-1-4-8-12/h1-10,14-15H,11H2/t14-,15-/m1/s1
Formel vikt 376.0758