Läs mer
2-Deoxy-2,2-difluoro-D-erythro-ribofuranose-3,5-dibenzoate, TRC
Produktkod. 30179072
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179072 500 mg
30179426 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179072 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD232785500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173824-58-2
Molekylformel C19 H16 F2 O6
Molekylvikt (g/mol) 378.32
Synonym [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-hydroxy-tetrahydrofuran-2-yl]methyl benzoate, D-erythro-Pentofuranose, 2-deoxy-2,2-difluoro-, 3,5-dibenzoate, 2-Deoxy-2,2-difluoro-D-erythro-ribofuranose-3,5-dibenzoate
IUPAC-namn [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-hydroxyoxolan-2-yl]methyl benzoate
LEDER OC1O[C@H](COC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)C1(F)F
Specifikationer
Kemiskt namn eller material 2-Deoxy-2,2-difluoro-D-erythro-ribofuranose-3,5-dibenzoate
Kvantitet 500 mg
InChI formel InChI=1S/C19H16F2O6/c20-19(21)15(27-17(23)13-9-5-2-6-10-13)14(26-18(19)24)11-25-16(22)12-7-3-1-4-8-12/h1-10,14-15,18,24H,11H2/t14-,15-,18?/m1/s1
Formel vikt 378.0915