Läs mer
2'-Deoxy-5-[3-[(2,2,2-trifluoroacetyl)amino]-1-propyn-1-yl]uridine, TRC
Produktkod. 30361210
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30361210 250 mg
1 options
Produktkod. 30361210 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD282515250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2'-Deoxy-5-[3-[(2,2,2-trifluoroacetyl)amino]-1-propyn-1-yl]uridine
CAS 115899-40-6
Molekylformel C14H14F3N3O6
Kvantitet 250 mg
Synonym 3: PN: US20060183135 FIGURE: 7 claimed sequence, 2'-Deoxy-5-[3-[(trifluoroacetyl)amino]-1-propynyl]uridine
InChI formel InChI=1S/C14H14F3N3O6/c15-14(16,17)12(24)18-3-1-2-7-5-20(13(25)19-11(7)23)10-4-8(22)9(6-21)26-10/h5,8-10,21-22H,3-4,6H2,(H,18,24)(H,19,23,25)/t8-,9+,10+/m0/s1
LEDER CS(=O)(=O)SCCN1CCN(CC\C=C2c3c(ccc(c3)C(F)(F)F)Sc4c/2cccc4)CC1
Molekylvikt (g/mol) 377.27