missing translation for 'onlineSavingsMsg'
Läs mer

2-Deoxystreptamine, Dihydrobromide, TRC

Produktkod. 17954915
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17954915 50 mg
17942695 100 mg
17933625 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17954915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD26350050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 84107-26-6
Molekylformel C6 H14 N2 O3 . 2 Br H
Molekylvikt (g/mol) 324.01
Synonym Streptamine, 2-deoxy-, hydrobromide (1:2), Streptamine, 2-deoxy-, dihydrobromide (9CI), 2-Deoxystreptamine dihydrobromide, 2-Deoxy-D-streptamine dihydrobromide
IUPAC-namn (1S,3R,4S,6R)-4,6-diaminocyclohexane-1,2,3-triol;dihydrobromide
LEDER Br.Br.N[C@@H]1C[C@H](N)[C@@H](O)[C@H](O)[C@H]1O

Specifikationer

Kemiskt namn eller material 2-Deoxystreptamine, Dihydrobromide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C6H14N2O3.2BrH/c7-2-1-3(8)5(10)6(11)4(2)9;;/h2-6,9-11H,1,7-8H2;2*1H/t2-,3+,4+,5-,6-;;
Formel vikt 321.9528
Procent renhet >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.