Läs mer
2’-Des(1-hydroxy-1-methylethyl)-2’-methycarboxy Montelukast, TRC
Produktkod. 30356815
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356815 100 mg
30355804 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356815 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD290040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2'-Des(1-hydroxy-1-methylethyl)-2'-methycarboxy Montelukast
CAS 851755-56-1
Molekylformel C34 H32 Cl N O4 S
Kvantitet 100 mg
Synonym 2-[(3R)-3-[[[1-(Carboxymethyl)cyclopropyl]methyl]thio]-3-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]propyl]-benzoic acid 1-methyl ester, (R)-2-[1-[[[1-[3-[(E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(methoxycarbonyl)phenyl]propyl]thio]methyl]cyclopropyl]acetic acid, 2-[1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-(2-methoxycarbonylphenyl)propyl]thio]methyl]cyclopropyl]acetic acid, 2-[1-[[[(1R)-1-[3-[(1E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(methoxycarbonyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic acid, Montelukast Sodium Imp. H (EP)
InChI formel InChI=1S/C34H32ClNO4S/c1-40-33(39)29-8-3-2-6-24(29)12-16-31(41-22-34(17-18-34)21-32(37)38)26-7-4-5-23(19-26)9-14-28-15-11-25-10-13-27(35)20-30(25)36-28/h2-11,13-15,19-20,31H,12,16-18,21-22H2,1H3,(H,37,38)/b14-9+/t31-/m1/s1
LEDER COC(=O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3
IUPAC-namn 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-(2-methoxycarbonylphenyl)propyl]sulfanylmethyl]cyclopropyl]acetic acid
Molekylvikt (g/mol) 586.14
Formel vikt 585.1741