Läs mer
2-Deschloro Aripiprazole, TRC
Produktkod. 30007625
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007625 500 mg
17950635 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD282020500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 203395-82-8
Molekylformel C23 H28 Cl N3 O2
Molekylvikt (g/mol) 413.94
Synonym 2(1H)-Quinolinone, 7-[4-[4-(3-chlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-, 2-Deschloro Aripiprazole, Aripiprazol Imp. D (EP), 7-[4-[4-(3-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one
IUPAC-namn 7-[4-[4-(3-chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
LEDER Clc1cccc(c1)N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2
Specifikationer
Kemiskt namn eller material 2-Deschloro Aripiprazole
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C23H28ClN3O2/c24-19-4-3-5-20(16-19)27-13-11-26(12-14-27)10-1-2-15-29-21-8-6-18-7-9-23(28)25-22(18)17-21/h3-6,8,16-17H,1-2,7,9-15H2,(H,25,28)
Formel vikt 413.187
Procent renhet >95%