Läs mer
2-Desethoxy-2-hydroxy-1H-1-Ethyl Candesartan Cilexetil, TRC
Produktkod. 30135271
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135271 25 mg
30135763 50 mg
18003340 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135271 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28875025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1185255-99-5
Molekylformel C33 H34 N6 O6
Molekylvikt (g/mol) 610.66
Synonym 1H-Benzimidazole-4-carboxylic acid, 3-[[2'-(1-ethyl-1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-2,3-dihydro-2-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, Candesartan Cilexetil Imp. C (EP)
IUPAC-namn 1-cyclohexyloxycarbonyloxyethyl 3-[[4-[2-(1-ethyltetrazol-5-yl)phenyl]phenyl]methyl]-2-oxo-1H-benzimidazole-4-carboxylate
LEDER CCn1nnnc1c2ccccc2c3ccc(CN4C(=O)Nc5cccc(C(=O)OC(C)OC(=O)OC6CCCCC6)c45)cc3
Specifikationer
Kemiskt namn eller material 2-Desethoxy-2-hydroxy-1H-1-Ethyl Candesartan Cilexetil
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C33H34N6O6/c1-3-39-30(35-36-37-39)26-13-8-7-12-25(26)23-18-16-22(17-19-23)20-38-29-27(14-9-15-28(29)34-32(38)41)31(40)43-21(2)44-33(42)45-24-10-5-4-6-11-24/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,34,41)
Formel vikt 610.254
Procent renhet >95%