missing translation for 'onlineSavingsMsg'
Läs mer

2-Desethoxy-2-hydroxy-1H-1-Ethyl Candesartan Cilexetil, TRC

Produktkod. 30135763
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135763 50 mg
30135271 25 mg
18003340 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30135763 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28875050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1185255-99-5
Molekylformel C33 H34 N6 O6
Molekylvikt (g/mol) 610.66
Synonym 1H-Benzimidazole-4-carboxylic acid, 3-[[2'-(1-ethyl-1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-2,3-dihydro-2-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, Candesartan Cilexetil Imp. C (EP)
IUPAC-namn 1-cyclohexyloxycarbonyloxyethyl 3-[[4-[2-(1-ethyltetrazol-5-yl)phenyl]phenyl]methyl]-2-oxo-1H-benzimidazole-4-carboxylate
LEDER CCn1nnnc1c2ccccc2c3ccc(CN4C(=O)Nc5cccc(C(=O)OC(C)OC(=O)OC6CCCCC6)c45)cc3

Specifikationer

Kemiskt namn eller material 2-Desethoxy-2-hydroxy-1H-1-Ethyl Candesartan Cilexetil
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C33H34N6O6/c1-3-39-30(35-36-37-39)26-13-8-7-12-25(26)23-18-16-22(17-19-23)20-38-29-27(14-9-15-28(29)34-32(38)41)31(40)43-21(2)44-33(42)45-24-10-5-4-6-11-24/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,34,41)
Formel vikt 610.254
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.