Läs mer
2-Desmethyl-2-hydroxymethyl Kresoxim-methyl Carboxylic Acid, TRC
Produktkod. 30366935
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366935 10 mg
30358864 1 mg
30349824 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28219510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1639810-41-5
Molekylformel C17 H17 N O5
Molekylvikt (g/mol) 315.32
Synonym Benzeneacetic acid, 2-[[2-(hydroxymethyl)phenoxy]methyl]-α-(methoxyimino)-, (αE)-, (αE)-2-[[2-(Hydroxymethyl)phenoxy]methyl]-α-(methoxyimino)benzeneacetic acid, BF 490-2
IUPAC-namn (2E)-2-[2-[[2-(hydroxymethyl)phenoxy]methyl]phenyl]-2-methoxyiminoacetic acid
LEDER CO\N=C(\C(=O)O)/c1ccccc1COc2ccccc2CO
Specifikationer
Kemiskt namn eller material (E)-Kresoxim-2-hydroxymethyl (free acid)
Kvantitet 10 mg
InChI formel InChI=1S/C17H17NO5/c1-22-18-16(17(20)21)14-8-4-2-7-13(14)11-23-15-9-5-3-6-12(15)10-19/h2-9,19H,10-11H2,1H3,(H,20,21)/b18-16+
Formel vikt 315.1107