Läs mer
2-Desmethyltetrahydrofuran-methylnaphthalene Nafronyl, TRC
Produktkod. 30355273
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30355273 10 mg
1 options
Produktkod. 30355273 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29391010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Desmethyltetrahydrofuran-methylnaphthalene Nafronyl
CAS 115025-98-4
Molekylformel C30H33NO2
Kvantitet 10 mg
Synonym alpha-(1-Naphthalenylmethyl)-1-naphthalenepropanoic Acid 2-(Diethylamino)ethyl Ester
InChI formel InChI=1S/C30H33NO2/c1-3-31(4-2)19-20-33-30(32)27(21-25-15-9-13-23-11-5-7-17-28(23)25)22-26-16-10-14-24-12-6-8-18-29(24)26/h5-18,27H,3-4,19-22H2,1-2H3
LEDER CCN(CC)CCOC(=O)C(Cc1cccc2ccccc12)Cc3cccc4ccccc34
Molekylvikt (g/mol) 439.59