Läs mer
2-(Dibenzylamino)-3',4'-dihydroxy-acetophenone, TRC
Produktkod. 30133283
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133283 100 mg
30134194 25 mg
17971185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133283 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD417520100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13062-58-3
Molekylformel C22H21NO3
Molekylvikt (g/mol) 347.41
Synonym N,N-Dibenzylarterenone, 2-[Bis(phenylmethyl)amino]-1-(3,4-dihydroxyphenyl)-ethanone;
IUPAC-namn 2-(dibenzylamino)-1-(3,4-dihydroxyphenyl)ethanone
LEDER OC1=C(O)C=CC(C(CN(CC2=CC=CC=C2)CC3=CC=CC=C3)=O)=C1
Specifikationer
Kemiskt namn eller material 2-(Dibenzylamino)-3',4'-dihydroxy-acetophenone
Kvantitet 100 mg
InChI formel InChI=1S/C22H21NO3/c24-20-12-11-19(13-21(20)25)22(26)16-23(14-17-7-3-1-4-8-17)15-18-9-5-2-6-10-18/h1-13,24-25H,14-16H2
Formel vikt 347.1521