Läs mer
2-(Diethylamino)ethanethiol Hydrochloride, TRC
Produktkod. 30373127
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30373127 10 g
1 options
Produktkod. 30373127 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD67995510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Diethylamino)ethanethiol Hydrochloride
CAS 1942-52-5
Molekylformel C6 H15 N S . Cl H
Kvantitet 10 g
Synonym Ethanethiol, 2-(diethylamino)-, hydrochloride (1:1), Ethanethiol, 2-(diethylamino)-, hydrochloride (6CI,8CI,9CI), (Diethylamino)ethanethiol hydrochloride, 2-(Diethylamino)ethanethiol hydrochloride, 2-(Diethylamino)ethylmercaptan hydrochloride, N,N-Diethyl
InChI formel InChI=1S/C6H15NS.ClH/c1-3-7(4-2)5-6-8;/h8H,3-6H2,1-2H3;1H
LEDER Cl.CCN(CC)CCS
IUPAC-namn 2-(diethylamino)ethanethiol;hydrochloride
Molekylvikt (g/mol) 169.716
Formel vikt 169.069