Läs mer
2-(Diethylamino)-N-(2,4-dimethylphenyl)acetamide Hydrochloride, Mikromol™
Produktkod. 30052063
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30052063 25 mg
1 options
Produktkod. 30052063 Leverantör Mikromol Leverantörsnummer MM0102.120025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(Diethylamino)-N-(2,4-dimethylphenyl)acetamide Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Diethylamino)-N-(2,4-dimethylphenyl)acetamide Hydrochloride
CAS 17289-54-2
Analyt- eller komponentnamn 2-(Diethylamino)-N-(2,4-dimethylphenyl)acetamide Hydrochloride
Frakt skick Room Temperature
Molekylformel C14 H22 N2 O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Lidocaine Hydrochloride Imp. I (EP), Lidocaine Imp. I (EP), 2-(Diethylamino)-N-(2,4-dimethylphenyl)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. I (EP) as Hydrochloride, Lidocaine Hydrochloride Impurity I as Hydrochloride, Lidocaine Impurity I as Hydrochloride
InChI formel InChI=1S/C14H22N2O.ClH/c1-5-16(6-2)10-14(17)15-13-8-7-11(3)9-12(13)4;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H
LEDER Cl.CCN(CC)CC(=O)Nc1ccc(C)cc1C
IUPAC-namn 2-(diethylamino)-N-(2,4-dimethylphenyl)acetamide;hydrochloride
Molekylvikt (g/mol) 270.8
Kvantitet 25 mg
Formel vikt 270.1499 g/mol