Läs mer
2-(Diethylamino)-N,N-diethylacetamide, TRC
Produktkod. 30369866
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30369866 100 mg
30380100 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30369866 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD443200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27794-54-3
Molekylformel C10H22N2O
Molekylvikt (g/mol) 186.29
Synonym N,N-Diethyl-2-(diethylamino)acetamide
LEDER CCN(CC)CC(=O)N(CC)CC
Specifikationer
Kemiskt namn eller material 2-(Diethylamino)-N,N-diethylacetamide
Kvantitet 100 mg
InChI formel InChI=1S/C10H22N2O/c1-5-11(6-2)9-10(13)12(7-3)8-4/h5-9H2,1-4H3
Formel vikt 186.1732