Läs mer
2-(Diethylazinoyl)-N-(2,6-dimethylphenyl)acetamide (Lidocaine N2-Oxide), Mikromol™
Produktkod. 18091930
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18091930 25 mg
18010540 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18091930 Leverantör Mikromol Leverantörsnummer MM0102.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(Diethylazinoyl)-N-(2,6-dimethylphenyl)acetamide (Lidocaine N2-Oxide)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lidocaine N-Oxide (2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide)
CAS 2903-45-9
Analyt- eller komponentnamn Lidocaine N-Oxide (2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide)
Frakt skick Ice pack (-20°C)
Molekylformel C14 H22 N2 O2
Certifikat och efterlevnad ISO 17025
Synonym Lidocaine Hydrochloride Imp. B (EP), Lidocaine Imp. B (EP), 2-(Diethylazinoyl)-N-(2,6-dimethylphenyl)acetamide, Lidocaine N2-Oxide, Lidocaine N-Oxide, 2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide, Lidocaine Hydrochloride Impurity B, Lidocaine Impurity B
InChI formel InChI=1S/C14H22N2O2/c1-5-16(18,6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)
LEDER CC[N+]([O-])(CC)CC(=O)Nc1c(C)cccc1C
IUPAC-namn 2-(2,6-dimethylanilino)-N,N-diethyl-2-oxoethanamine oxide
Molekylvikt (g/mol) 250.34
Kvantitet 25 mg
Formel vikt 250.1681 g/mol