Learn More
2-(Difluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
Catalog Number p-7027485
Click to view available options
Quantity:
1 g
5 g
71653-64-0
C8H6F2O2
172.131
MFCD00042251
QPBNHDFPMRENBC-UHFFFAOYSA-N
2-difluoromethoxy benzaldehyde, 2-difluoromethoxy-benzaldehyde, o-difluoromethoxybenzaldehyde, 2-difloromethoxybenzaldehyde, pubchem2906, acmc-209ojk, difluoromethoxybenzaldehyde, ksc356m9l, zerenex zx009675, benzaldehyde, difluoromethoxy
2736984
2-(difluoromethoxy)benzaldehyde
C1=CC=C(C(=C1)C=O)OC(F)F
71653-64-0
C8H6F2O2
172.131
MFCD00042251
QPBNHDFPMRENBC-UHFFFAOYSA-N
2-difluoromethoxy benzaldehyde, 2-difluoromethoxy-benzaldehyde, o-difluoromethoxybenzaldehyde, 2-difloromethoxybenzaldehyde, pubchem2906, acmc-209ojk, difluoromethoxybenzaldehyde, ksc356m9l, zerenex zx009675, benzaldehyde, difluoromethoxy
2736984
2-(difluoromethoxy)benzaldehyde
C1=CC=C(C(=C1)C=O)OC(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-(Difluoromethoxy)benzaldehyde is an important raw material used as an intermediate in the preparation of pharmaceuticals, agrochemicals, and dyestuffs. It is also involved in the preparation of 4-(2-difluoromethoxyphenyl)- substituted 1,4-dihydropyridine 3,5-diesters.
Solubility
Miscible with 2-propanol and acetic acid.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.284
Boiling Point 88°C to 90°C (9 mmHg)
Flash Point 106°C (223°F)
Refractive Index 1.497
Quantity 1 g
Beilstein 389068
Sensitivity Air Sensitive
Solubility Information Miscible with 2-propanol and acetic acid.
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde