Läs mer
2-(Difluoromethoxy)pyridin-4-amine, TRC
Produktkod. 30373712
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373712 10 mg
30370856 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30373712 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45275310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Difluoromethoxy)pyridin-4-amine
CAS 1211587-46-0
Molekylformel C6H6N2OF2
Kvantitet 10 mg
InChI formel InChI=1S/C6H6F2N2O/c7-6(8)11-5-3-4(9)1-2-10-5/h1-3,6H,(H2,9,10)
LEDER Nc1ccnc(OC(F)F)c1
Molekylvikt (g/mol) 160.12
Formel vikt 160.0448