Learn More
2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Thermo Scientific™
Catalog Number p-4812651
Click to view available options
Quantity:
1 g
5 g
195886-79-4
C7H4F4
164.1
FDUVCOVVYFCQEH-UHFFFAOYSA-N
2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
46311703
2-(difluoromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)C(F)F)F
195886-79-4
C7H4F4
164.1
FDUVCOVVYFCQEH-UHFFFAOYSA-N
2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
46311703
2-(difluoromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)C(F)F)F
Boiling Point 120°C
Flash Point 28°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.1
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethyl)-1, 4-difluorobenzene
compliance-icons compliance-icons
Product Identifier
  • 2-(Difluoromethyl)-1,4-difluorobenzene, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.